siRNA Details
Gene Symbol: IGF2BP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007225.1 | NP_001007226.1 | 15 | 2033 | ||
NM_001291869.1 | NP_001278798.1 | 16 | 2180 | |||
NM_001291872.1 | NP_001278801.1 | 15 | 1982 | |||
NM_001291873.1 | NP_001278802.1 | 15 | 1964 | |||
NM_001291874.1 | NP_001278803.1 | 14 | 1835 | |||
NM_001291875.1 | NP_001278804.1 | 14 | 1900 | |||
NM_006548.4 | NP_006539.3 | 16 | 2162 | |||
XM_006713477.3 | XP_006713540.1 | 11 | 1586 | |||
XM_006713478.3 | XP_006713541.1 | 10 | 1452 | |||
XM_017005557.1 | XP_016861046.1 | 16 | 2159 | |||
XM_017005558.1 | XP_016861047.1 | 15 | 2056 | |||
XM_017005559.1 | XP_016861048.1 | 15 | 2054 | |||
XM_017005560.1 | XP_016861049.1 | 16 | 2082 | |||
GenBank mRNA | AK296703.1 | - | 10 | 1297 | ||
BC021290.2 | - | 16 | 2147 | 5090334 | ||
BX537583.1 | - | 1 | 1371 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
