siRNA Details
Gene Symbol: CCDC144B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_036647.1 | 17 | 5565 | |||
GenBank mRNA | BX647826.1 | - | 17 | 5556 |
Gene Symbol: CALU | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130674.2 | NP_001124146.1 | 7 | 5270 | ||
NM_001199671.1 | NP_001186600.1 | 8 | 6085 | |||
NM_001199672.1 | NP_001186601.1 | 8 | 6085 | |||
NM_001199673.1 | NP_001186602.1 | 6 | 5070 | |||
NM_001219.4 | NP_001210.1 | 7 | 5270 | |||
NR_074086.1 | 5 | 4844 |
Gene Symbol: CCDC144A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014695.2 | NP_055510.1 | 18 | 5583 | ||
NR_130142.1 | 16 | 5298 | ||||
GenBank mRNA | BC133019.1 | - | 18 | 5583 | 40147041 | |
BC144224.1 | - | 16 | 5297 | 9052744 | ||
BC144225.1 | - | 18 | 5583 | 9052745 |
Gene Symbol: DSC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001941.4 | NP_001932.2 | 16 | 7134 | ||
NM_024423.3 | NP_077741.2 | 17 | 7177 |
Gene Symbol: GBP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052941.4 | NP_443173.2 | 11 | 2476 | ||
GenBank mRNA | AK131094.1 | - | 11 | 2389 | ||
AL832576.2 | - | 11 | 2407 | |||
BC070055.1 | - | 11 | 2413 | 30345441 |
Gene Symbol: LINC00670 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_034144.1 | 3 | 1363 | |||
NR_034145.1 | 4 | 1525 | ||||
GenBank mRNA | AK092009.1 | - | 4 | 1526 |
Gene Symbol: LINC00891 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK128305.1 | - | 3 | 1139 |
Gene Symbol: LOC101929163 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK097727.1 | - | 1 | 841 | ||
AK123889.1 | - | 2 | 1642 |
Gene Symbol: OR11A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013937.3 | NP_039225.1 | 1 | 1238 | ||
GenBank mRNA | AJ459857.1 | - | 1 | 1238 |
Gene Symbol: RAP1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK127392.1 | - | 2 | 570 |
Gene Symbol: SIGLEC14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017026113.1 | XP_016881602.1 | 6 | 2448 |
Gene Symbol: SLC35D2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_104627.1 | 13 | 1533 | |||
GenBank mRNA | AK024841.1 | - | 2 | 516 | ||
AK307890.1 | - | 13 | 1513 |
Gene Symbol: SLC4A5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB209752.1 | - | 1 | 365 |
Gene Symbol: UBAC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BX647463.1 | - | 1 | 824 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
