siRNA Details
Gene Symbol: THAP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008695.1 | NP_001008695.1 | 5 | 1055 | ||
NM_030573.2 | NP_085050.2 | 4 | 1229 | |||
GenBank mRNA | AK024609.1 | - | 2 | 1796 | ||
AK025906.1 | - | 2 | 1561 | |||
AK300715.1 | - | 5 | 844 | |||
BC004346.1 | - | 4 | 1200 | 3633743 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
