siRNA Details
Gene Symbol: ATN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007026.1 | NP_001007027.1 | 10 | 4150 | ||
NM_001940.3 | NP_001931.2 | 10 | 4143 | |||
GenBank mRNA | BC051795.2 | - | 10 | 4164 | 4181592 | |
D31840.1 | - | 10 | 4136 | |||
D38529.1 | - | 9 | 4055 | |||
U23851.1 | - | 9 | 3968 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
