siRNA Details
Gene Symbol: RWDD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007464.2 | NP_001007465.1 | 4 | 487 | ||
NM_015952.3 | NP_057036.2 | 3 | 373 | |||
NM_016104.3 | NP_057188.2 | 5 | 615 | |||
GenBank mRNA | AF092134.1 | - | 3 | 330 | ||
AF132958.1 | - | 3 | 243 | |||
AK290727.1 | - | 3 | 373 | |||
AK301089.1 | - | 4 | 350 | |||
BC015802.1 | - | 3 | 229 | 3909348 | ||
CF128570.1 | - | 3 | 320 | |||
CN363729.1 | - | 5 | 456 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
