siRNA Details
Gene Symbol: C1QTNF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030945.3 | NP_112207.1 | 6 | 2970 | ||
NM_181435.5 | NP_852100.3 | 6 | 3189 | |||
GenBank mRNA | BX640995.1 | - | 7 | 2790 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
