siRNA Details
Gene Symbol: MTX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006635.2 | NP_001006636.1 | 10, 11 | 1068 | ||
NM_001319097.1 | NP_001306026.1 | 9, 10 | 795 | |||
NM_006554.4 | NP_006545.1 | 9, 10 | 864 | |||
XM_011510497.2 | XP_011508799.1 | 10, 11 | 961 | |||
GenBank mRNA | AF053551.1 | - | 9, 10 | 837 | ||
AK303564.1 | - | 10, 11 | 993 | |||
AK315918.1 | - | 10, 11 | 1004 | |||
BC017271.2 | - | 10, 11 | 998 | 4635884 | ||
BC067831.1 | - | 9, 10 | 843 | 5298149 | ||
BC088359.1 | - | 9, 10 | 799 | 30378040 | ||
CX165048.1 | - | 9, 10 | 688 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
