siRNA Details
Gene Symbol: GEMIN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009182.1 | NP_001009182.1 | 4 | 406 | ||
NM_001009183.1 | NP_001009183.1 | 4 | 406 | |||
NM_003616.2 | NP_003607.1 | 4 | 406 | |||
XM_017021709.1 | XP_016877198.1 | 4 | 429 | |||
GenBank mRNA | AB037701.1 | - | 4 | 384 | ||
AB037702.1 | - | 4 | 384 | |||
AB037703.1 | - | 4 | 304 | |||
AF027150.1 | - | 4 | 467 | |||
AK313947.1 | - | 4 | 394 | |||
BC028095.1 | - | 4 | 405 | |||
BC104968.1 | - | 4 | 405 | 8143971 | ||
CN484054.1 | - | 4 | 406 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
