siRNA Details
Gene Symbol: MICB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289160.1 | NP_001276089.1 | 6 | 2245 | ||
NM_001289161.1 | NP_001276090.1 | 6 | 2215 | |||
NM_005931.4 | NP_005922.2 | 6 | 2344 | |||
GenBank mRNA | BC044218.1 | - | 6 | 2177 | 6025253 | |
X91625.1 | - | 6 | 2201 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
