siRNA Details
Gene Symbol: DPH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039589.1 | NP_001034678.1 | 2 | 416 | ||
NM_001319165.1 | NP_001306094.1 | 2 | 406 | |||
NM_001319166.1 | NP_001306095.1 | 2 | 416 | |||
NM_001319167.1 | NP_001306096.1 | 2 | 364 | |||
NM_001319168.1 | NP_001306097.1 | 2 | 364 | |||
NM_001319169.1 | NP_001306098.1 | 2 | 416 | |||
NM_001319170.1 | NP_001306099.1 | 2 | 416 | |||
NM_001319171.1 | NP_001306100.1 | 2 | 416 | |||
NM_001384.4 | NP_001375.2 | 2 | 416 | |||
XM_017000502.1 | XP_016855991.1 | 2 | 418 | |||
XM_017000503.1 | XP_016855992.1 | 2 | 352 | |||
XM_017000504.1 | XP_016855993.1 | 2 | 400 | |||
XM_017000505.1 | XP_016855994.1 | 2 | 332 | |||
XM_017000506.1 | XP_016855995.1 | 2 | 400 | |||
GenBank mRNA | AF053003.1 | - | 2 | 382 | ||
AK091303.1 | - | 2 | 335 | |||
AK297933.1 | - | 2 | 389 | |||
AK315506.1 | - | 2 | 400 | |||
BC001389.2 | - | 2 | 375 | 3051455 | ||
BC003181.1 | - | 2 | 393 | 2958202 | ||
BC016956.1 | - | 2 | 375 | 3900854 | ||
BG385934.1 | - | 2 | 401 | |||
BG472282.1 | - | 2 | 374 | |||
BG747150.1 | - | 2 | 344 | |||
BM564151.1 | - | 367 | ||||
BM794530.1 | - | 2 | 358 | |||
BQ056213.1 | - | 2 | 390 | |||
BQ215136.1 | - | 2 | 424 | |||
BT007431.1 | - | 2 | 221 | |||
CX164803.1 | - | 2 | 343 | |||
DA906783.1 | - | 2 | 418 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
