siRNA Details
Gene Symbol: TPT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286272.1 | NP_001273201.1 | 6 | 971 | ||
NM_001286273.1 | NP_001273202.1 | 5 | 732 | |||
NM_003295.3 | NP_003286.1 | 6 | 806 | |||
GenBank mRNA | AK130847.1 | - | 6 | 705 | ||
AK296587.1 | - | 6 | 615 | |||
AY117678.1 | - | 6 | 706 | |||
BC003352.2 | - | 6 | 689 | 2899964 | ||
BC012431.1 | - | 6 | 665 | 3880559 | ||
BC022436.1 | - | 5 | 660 | 4281364 | ||
BC040008.1 | - | 3 | 1712 | |||
BC052333.1 | - | 6 | 695 | 6058799 | ||
L13806.1 | - | 4 | 426 | |||
X16064.1 | - | 6 | 685 | |||
X64899.1 | - | 6 | 677 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
