siRNA Details
Gene Symbol: TCEAL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006935.2 | NP_001006936.1 | 3 | 656 | ||
NM_001006937.2 | NP_001006938.1 | 3 | 651 | |||
NM_001300901.1 | NP_001287830.1 | 5 | 1084 | |||
NM_001305840.1 | NP_001292769.1 | 4 | 905 | |||
NM_001305841.1 | NP_001292770.1 | 2 | 578 | |||
NM_001305842.1 | NP_001292771.1 | 3 | 710 | |||
NM_024863.5 | NP_079139.4 | 3 | 715 | |||
XM_017029856.1 | XP_016885345.1 | 3 | 975 | |||
XM_017029857.1 | XP_016885346.1 | 3 | 986 | |||
GenBank mRNA | AF271783.1 | - | 3 | 628 | ||
AF314542.1 | - | 5 | 1084 | |||
AK024827.1 | - | 3 | 649 | |||
AK093021.1 | - | 1 | 1582 | |||
AK225117.1 | - | 3 | 649 | |||
AL833640.1 | - | 3 | 709 | |||
AY885169.1 | - | 4 | 532 | |||
BC012296.1 | - | 3 | 615 | 3445403 | ||
BI084326.1 | - | 2 | 549 | |||
BX104938.1 | - | 1 | 103 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
