siRNA Details
Gene Symbol: CBWD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172003.3 | NP_742000.1 | 6, 7 | 717 | ||
XM_005263600.2 | XP_005263657.1 | 6, 7 | 717 | |||
XM_005263602.2 | XP_005263659.1 | 6, 7 | 717 | |||
XM_005263603.2 | XP_005263660.1 | 7, 8 | 751 | |||
XM_005263604.2 | XP_005263661.1 | 6, 7 | 717 | |||
XM_005263605.3 | XP_005263662.1 | 7, 8 | 752 | |||
XM_011510667.1 | XP_011508969.1 | 6, 7 | 717 | |||
XM_011510668.2 | XP_011508970.1 | 3, 4 | 277 | |||
XM_011510669.2 | XP_011508971.1 | 7, 8 | 761 | |||
XM_017003407.1 | XP_016858896.1 | 6, 7 | 717 | |||
XM_017003408.1 | XP_016858897.1 | 8, 9 | 881 | |||
GenBank mRNA | AF452722.1 | - | 6, 7 | 573 | ||
AK225251.1 | - | 7, 8 | 731 | |||
AK290072.1 | - | 6, 7 | 717 | |||
AK303653.1 | - | 7, 8 | 756 | |||
AY343910.1 | - | 6, 7 | 602 | |||
BC120991.1 | - | 6, 7 | 588 | 40116409 | ||
BC120992.1 | - | 6, 7 | 588 | 40116411 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
