siRNA Details
Gene Symbol: DNAJC15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013238.2 | NP_037370.2 | 4 | 695 | ||
GenBank mRNA | AF126743.1 | - | 5 | 717 | ||
AJ270691.1 | - | 1 | 61 | |||
AK311866.1 | - | 4 | 331 | |||
AY248898.1 | - | 4 | 695 | |||
AY512565.1 | - | 4 | 388 | |||
BC010910.1 | - | 4 | 424 | 4332619 | ||
BC095400.1 | - | 4 | 314 | 30530999 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
