siRNA Details
Gene Symbol: DNAJC15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013238.2 | NP_037370.2 | 6 | 966 | ||
GenBank mRNA | AF126743.1 | - | 7 | 988 | ||
BC010910.1 | - | 6 | 695 | 4332619 | ||
BC095400.1 | - | 6 | 587 | 30530999 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
