siRNA Details
Gene Symbol: MMD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012329.2 | NP_036461.2 | 7 | 1233 | ||
XM_006721795.2 | XP_006721858.1 | 8 | 1339 | |||
GenBank mRNA | AY424289.1 | - | 7 | 1213 | ||
BC026324.1 | - | 7 | 1205 | 4794375 | ||
X85750.1 | - | 7 | 1030 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
