siRNA Details
Gene Symbol: MMD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012329.2 | NP_036461.2 | 7 | 2572 | ||
XM_006721795.2 | XP_006721858.1 | 8 | 2678 | |||
GenBank mRNA | AY424289.1 | - | 7 | 2551 | ||
BC026324.1 | - | 7 | 2542 | 4794375 | ||
X85750.1 | - | 7 | 2368 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
