siRNA Details
Gene Symbol: OVCA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080822.2 | NP_543012.1 | 2 | 889 | ||
GenBank mRNA | AF073528.1 | - | 2 | 824 | ||
AF321875.1 | - | 2 | 824 | |||
BC040696.2 | - | 2 | 839 | 6158258 | ||
BC041170.1 | - | 2 | 839 | 5738727 |
Gene Symbol: DPH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001383.3 | NP_001374.3 | 13 | 2008 | ||
GenBank mRNA | AF321876.1 | - | 13 | 2008 | ||
AK090530.1 | - | 7 | 2081 | |||
AK098268.1 | - | 2 | 2428 | |||
BC003099.1 | - | 13 | 1970 | |||
S81752.1 | - | 13 | 2043 | |||
U34880.1 | - | 13 | 2043 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
