siRNA Details
Gene Symbol: TFIP11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008697.1 | NP_001008697.1 | 15 | 2347 | ||
NM_012143.2 | NP_036275.1 | 14 | 2270 | |||
XM_011530079.1 | XP_011528381.1 | 15 | 2330 | |||
XM_017028735.1 | XP_016884224.1 | 15 | 2488 | |||
GenBank mRNA | AF070662.1 | - | 1 | 343 | ||
AK025443.1 | - | 13 | 2394 | |||
AK025739.1 | - | 10 | 1728 | |||
AK298303.1 | - | 10 | 1330 | |||
AK299964.1 | - | 9 | 1096 | |||
AL050258.1 | - | 14 | 2283 | |||
AL080147.1 | - | 14 | 2250 | |||
BC011599.2 | - | 14 | 2247 | 3051032 | ||
BC033080.1 | - | 14 | 2248 | 3544615 | ||
BT007274.1 | - | 11 | 2020 | |||
CT841511.2 | - | 12 | 2122 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
