siRNA Details
Gene Symbol: TFIP11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008697.1 | NP_001008697.1 | 6 | 652 | ||
NM_012143.2 | NP_036275.1 | 5 | 575 | |||
XM_011530079.1 | XP_011528381.1 | 6 | 635 | |||
XM_017028735.1 | XP_016884224.1 | 6 | 793 | |||
GenBank mRNA | AK025443.1 | - | 4 | 699 | ||
AK025739.1 | - | 1 | 33 | |||
AK299964.1 | - | 5 | 547 | |||
AL050258.1 | - | 5 | 588 | |||
AL080147.1 | - | 5 | 555 | |||
BC011599.2 | - | 5 | 552 | 3051032 | ||
BC033080.1 | - | 5 | 553 | 3544615 | ||
BT007274.1 | - | 2 | 325 | |||
BX354919.2 | - | 6 | 651 | |||
CT841511.2 | - | 3 | 427 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
