siRNA Details
Gene Symbol: TFIP11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008697.1 | NP_001008697.1 | 7, 8 | 852 | ||
NM_012143.2 | NP_036275.1 | 6, 7 | 775 | |||
XM_011530079.1 | XP_011528381.1 | 7, 8 | 835 | |||
XM_017028735.1 | XP_016884224.1 | 7, 8 | 993 | |||
GenBank mRNA | AK025443.1 | - | 5, 6 | 899 | ||
AK025739.1 | - | 2, 3 | 233 | |||
AL050258.1 | - | 6, 7 | 788 | |||
AL080147.1 | - | 6, 7 | 755 | |||
BC011599.2 | - | 6, 7 | 752 | 3051032 | ||
BC033080.1 | - | 6, 7 | 753 | 3544615 | ||
BT007274.1 | - | 3, 4 | 525 | |||
BX354919.2 | - | 7, 8 | 852 | |||
CT841511.2 | - | 4, 5 | 627 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
