siRNA Details
Gene Symbol: MTIF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005369.1 | NP_001005369.1 | 5 | 799 | ||
NM_001321001.1 | NP_001307930.1 | 4 | 334 | |||
NM_001321002.1 | NP_001307931.1 | 4 | 306 | |||
NM_001321003.1 | NP_001307932.1 | 5 | 516 | |||
NM_001321004.1 | NP_001307933.1 | 3 | 568 | |||
NM_001321005.1 | NP_001307934.1 | 4 | 378 | |||
NM_002453.2 | NP_002444.2 | 4 | 378 | |||
XM_005264335.3 | XP_005264392.1 | 2 | 177 | |||
XM_011532871.2 | XP_011531173.1 | 3 | 211 | |||
XM_017004162.1 | XP_016859651.1 | 5 | 507 | |||
XM_017004163.1 | XP_016859652.1 | 4 | 583 | |||
XM_017004164.1 | XP_016859653.1 | 4 | 372 | |||
XM_017004165.1 | XP_016859654.1 | 5 | 571 | |||
XM_017004166.1 | XP_016859655.1 | 5 | 571 | |||
GenBank mRNA | AF494407.1 | - | 4 | 358 | ||
AK225184.1 | - | 4 | 347 | |||
AK307993.1 | - | 4 | 358 | |||
BC017243.1 | - | 4 | 326 | |||
BC039848.1 | - | 4 | 336 | |||
BC054048.1 | - | 4 | 275 | |||
BC064977.1 | - | 4 | 318 | |||
BC067352.1 | - | 5 | 799 | |||
BP282053.1 | - | 4 | 286 | |||
CB961193.1 | - | 4 | 405 | |||
DB478758.2 | - | 3 | 537 | |||
L34600.1 | - | 4 | 332 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
