siRNA Details
Gene Symbol: ERCC8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000082.3 | NP_000073.1 | 4 | 452 | ||
NM_001007233.2 | NP_001007234.1 | 5 | 670 | |||
NM_001007234.2 | NP_001007235.1 | 4 | 452 | |||
NM_001290285.1 | NP_001277214.1 | 4 | 452 | |||
GenBank mRNA | AK056931.1 | - | 5 | 643 | ||
AK290726.1 | - | 4 | 432 | |||
AK294856.1 | - | 4 | 433 | |||
AK304610.1 | - | 4 | 382 | |||
AK314511.1 | - | 4 | 425 | |||
AL691658.1 | - | 4 | 439 | |||
BC009793.1 | - | 4 | 429 | 3683434 | ||
BT020021.1 | - | 4 | 382 | |||
CR536563.1 | - | 4 | 382 | |||
DA169067.1 | - | 4 | 452 | |||
KU177979.1 | - | 4 | 382 | |||
KU177980.1 | - | 2 | 184 | |||
U28413.1 | - | 4 | 418 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
