siRNA Details
Gene Symbol: ERCC8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000082.3 | NP_000073.1 | 6 | 607 | ||
NM_001007233.2 | NP_001007234.1 | 7 | 825 | |||
NM_001007234.2 | NP_001007235.1 | 6 | 607 | |||
NM_001290285.1 | NP_001277214.1 | 5 | 525 | |||
GenBank mRNA | AK056931.1 | - | 7 | 798 | ||
AK226129.1 | - | 3 | 384 | |||
AK290726.1 | - | 6 | 587 | |||
AK294856.1 | - | 5 | 506 | |||
AK304610.1 | - | 6 | 537 | |||
AK314511.1 | - | 6 | 580 | |||
BC009793.1 | - | 6 | 584 | 3683434 | ||
BT020021.1 | - | 6 | 537 | |||
CR536563.1 | - | 6 | 537 | |||
KU177979.1 | - | 6 | 537 | |||
KU177980.1 | - | 4 | 343 | |||
U28413.1 | - | 6 | 573 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
