siRNA Details
Gene Symbol: WDR89 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008726.2 | NP_001008726.1 | 4 | 564 | ||
NM_001258272.1 | NP_001245201.1 | 3 | 960 | |||
NM_080666.3 | NP_542397.1 | 3 | 455 | |||
XM_011536383.2 | XP_011534685.1 | 2 | 494 | |||
XM_011536385.2 | XP_011534687.1 | 4 | 462 | |||
GenBank mRNA | AF115513.1 | - | 3 | 387 | ||
BC010698.1 | - | 2 | 347 | 3870073 | ||
BI603622.1 | - | 3 | 411 | |||
BP278550.1 | - | 4 | 475 | |||
BX248008.1 | - | 3 | 444 | |||
DA882425.1 | - | 3 | 456 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
