siRNA Details
Gene Symbol: GEMIN7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007269.1 | NP_001007270.1 | 2 | 1405 | ||
NM_001007270.1 | NP_001007271.1 | 3 | 1442 | |||
NM_024707.2 | NP_078983.1 | 3 | 1528 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
