siRNA Details
Gene Symbol: GEMIN7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007269.1 | NP_001007270.1 | 2 | 724 | ||
NM_001007270.1 | NP_001007271.1 | 3 | 761 | |||
NM_001319054.1 | NP_001305983.1 | 2 | 703 | |||
NM_001319055.1 | NP_001305984.1 | 2 | 750 | |||
NM_024707.2 | NP_078983.1 | 3 | 847 | |||
XM_005259262.3 | XP_005259319.1 | 3 | 975 | |||
XM_005259263.4 | XP_005259320.1 | 2 | 713 | |||
XM_017027312.1 | XP_016882801.1 | 2 | 950 | |||
GenBank mRNA | AK024018.1 | - | 3 | 831 | ||
BC007793.2 | - | 3 | 819 | 4053402 | ||
BM009097.1 | - | 2 | 705 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
