siRNA Details
Gene Symbol: ADAMTSL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001288607.1 | NP_001275536.1 | 20 | 3276 | ||
NM_001288608.1 | NP_001275537.1 | 20 | 3462 | |||
NM_019032.5 | NP_061905.2 | 19 | 3393 | |||
XM_011509644.2 | XP_011507946.1 | 19 | 3424 | |||
XM_011509645.2 | XP_011507947.1 | 18 | 3352 | |||
XM_011509648.2 | XP_011507950.1 | 19 | 3378 | |||
XM_011509651.2 | XP_011507953.1 | 13 | 2174 | |||
XM_011509652.1 | XP_011507954.1 | 13 | 1880 | |||
XM_017001506.1 | XP_016856995.1 | 20 | 3833 | |||
XM_017001507.1 | XP_016856996.1 | 11 | 1615 | |||
GenBank mRNA | AF217974.1 | - | 8 | 1194 | ||
AF370411.1 | - | 13 | 1930 | |||
AK293037.1 | - | 9 | 1325 | |||
AL122085.1 | - | 3 | 216 | |||
BC027478.1 | - | 12 | 1832 | |||
BC071852.1 | - | 11 | 1689 | |||
BC094811.1 | - | 11 | 1689 | |||
BC140800.1 | - | 19 | 3314 | 9021668 | ||
BC144581.1 | - | 19 | 3128 | 9053111 | ||
BX647294.1 | - | 6 | 955 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
