siRNA Details
Gene Symbol: ADAMTSL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001288607.1 | NP_001275536.1 | 18 | 3012 | ||
NM_001288608.1 | NP_001275537.1 | 18 | 3198 | |||
NM_019032.5 | NP_061905.2 | 17 | 3129 | |||
XM_011509644.2 | XP_011507946.1 | 17 | 3160 | |||
XM_011509645.2 | XP_011507947.1 | 16 | 3088 | |||
XM_011509648.2 | XP_011507950.1 | 17 | 3114 | |||
XM_011509651.2 | XP_011507953.1 | 11 | 1910 | |||
XM_011509652.1 | XP_011507954.1 | 11 | 1616 | |||
XM_017001506.1 | XP_016856995.1 | 18 | 3569 | |||
XM_017001507.1 | XP_016856996.1 | 9 | 1351 | |||
GenBank mRNA | AF217974.1 | - | 6 | 930 | ||
AF370411.1 | - | 11 | 1666 | |||
AK092509.1 | - | 8 | 1248 | |||
AK293037.1 | - | 7 | 1061 | |||
BC027478.1 | - | 10 | 1568 | |||
BC071852.1 | - | 9 | 1425 | |||
BC094811.1 | - | 9 | 1425 | |||
BC140800.1 | - | 17 | 3050 | 9021668 | ||
BC144581.1 | - | 17 | 2864 | 9053111 | ||
BX647294.1 | - | 4 | 691 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
