siRNA Details
Gene Symbol: ADAMTSL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001288607.1 | NP_001275536.1 | 18 | 2973 | ||
NM_001288608.1 | NP_001275537.1 | 18 | 3159 | |||
NM_019032.5 | NP_061905.2 | 17 | 3090 | |||
XM_011509644.2 | XP_011507946.1 | 17 | 3121 | |||
XM_011509645.2 | XP_011507947.1 | 16 | 3049 | |||
XM_011509648.2 | XP_011507950.1 | 17 | 3075 | |||
XM_011509651.2 | XP_011507953.1 | 11 | 1871 | |||
XM_011509652.1 | XP_011507954.1 | 11 | 1577 | |||
XM_017001506.1 | XP_016856995.1 | 18 | 3530 | |||
XM_017001507.1 | XP_016856996.1 | 9 | 1312 | |||
GenBank mRNA | AF217974.1 | - | 6 | 891 | ||
AF370411.1 | - | 11 | 1627 | |||
AK092509.1 | - | 8 | 1209 | |||
AK293037.1 | - | 7 | 1022 | |||
BC027478.1 | - | 10 | 1529 | |||
BC071852.1 | - | 9 | 1386 | |||
BC094811.1 | - | 9 | 1386 | |||
BC140800.1 | - | 17 | 3011 | 9021668 | ||
BC144581.1 | - | 17 | 2825 | 9053111 | ||
BX647294.1 | - | 4 | 652 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
