siRNA Details
Gene Symbol: TCTEX1D2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152773.4 | NP_689986.2 | 2 | 286 | ||
GenBank mRNA | BC015348.1 | - | 2 | 222 | ||
BC021177.2 | - | 2 | 266 | 4830500 | ||
BM548106.1 | - | 2 | 311 | |||
CK818899.1 | - | 2 | 280 |
Gene Symbol: TM4SF19-TCTEX1D2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037950.1 | 6 | 892 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
