siRNA Details
Gene Symbol: WDR35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006657.1 | NP_001006658.1 | 16 | 1699 | ||
NM_020779.3 | NP_065830.2 | 15 | 1666 | |||
XM_011533007.2 | XP_011531309.1 | 5 | 391 | |||
GenBank mRNA | AB037757.2 | - | 16 | 1707 | ||
AK091205.1 | - | 9 | 1034 | |||
AK122917.1 | - | 15 | 1591 | |||
AK315094.1 | - | 15 | 1591 | |||
AL832903.1 | - | 4 | 196 | |||
BC036659.2 | - | 15 | 1665 | 5266940 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
