siRNA Details
Gene Symbol: BOD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159651.1 | NP_001153123.1 | 3 | 952 | ||
NM_138369.2 | NP_612378.1 | 4 | 1149 | |||
XM_017010017.1 | XP_016865506.1 | 2 | 1241 | |||
GenBank mRNA | AK301982.1 | - | 4 | 1137 | ||
BC003114.1 | - | 3 | 877 | 3537973 | ||
BC007436.2 | - | 4 | 1139 | 2961368 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
