siRNA Details
Gene Symbol: GOLGA8A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181077.3 | NP_851422.1 | 8, 9 | 757 | ||
NR_027409.1 | 16, 17 | 3019 | ||||
XM_006720440.2 | XP_006720503.1 | 8, 9 | 725 | |||
XM_017022018.1 | XP_016877507.1 | 12, 13 | 3153 | |||
XM_017022019.1 | XP_016877508.1 | 11, 12 | 2849 | |||
XM_017022020.1 | XP_016877509.1 | 6, 7 | 847 | |||
GenBank mRNA | AB020662.1 | - | 7, 8 | 737 | ||
AF163441.1 | - | 6, 7 | 1181 | |||
AF204231.1 | - | 7, 8 | 1074 | |||
AL137686.1 | - | 6, 7 | 481 | |||
BC032774.1 | - | 8, 9 | 659 | |||
BC093673.1 | - | 8, 9 | 703 | 7939518 | ||
BC104800.1 | - | 8, 9 | 703 | 8143803 | ||
BC144609.1 | - | 6, 7 | 494 | 9053140 | ||
BC150329.1 | - | 7, 8 | 748 | |||
BC152410.1 | - | 7, 8 | 737 | |||
BX648160.1 | - | 15, 16 | 2973 | |||
BX648575.1 | - | 15, 16 | 2726 | |||
CR749361.1 | - | 8, 9 | 1910 |
Gene Symbol: GOLGA8B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001023567.4 | NP_001018861.3 | 8, 9 | 757 | ||
NR_027410.1 | 16, 17 | 3005 | ||||
XM_011521596.1 | XP_011519898.1 | 15, 16 | 1602 | |||
XM_011521597.1 | XP_011519899.1 | 14, 15 | 1553 | |||
XM_017022210.1 | XP_016877699.1 | 16, 17 | 1906 | |||
XM_017022211.1 | XP_016877700.1 | 16, 17 | 1675 | |||
XM_017022212.1 | XP_016877701.1 | 15, 16 | 2229 | |||
XM_017022213.1 | XP_016877702.1 | 15, 16 | 1631 | |||
XM_017022214.1 | XP_016877703.1 | 16, 17 | 1906 | |||
XM_017022215.1 | XP_016877704.1 | 16, 17 | 2428 | |||
XM_017022216.1 | XP_016877705.1 | 15, 16 | 3026 | |||
XM_017022217.1 | XP_016877706.1 | 14, 15 | 2948 | |||
XM_017022218.1 | XP_016877707.1 | 16, 17 | 3055 | |||
GenBank mRNA | AK097551.1 | - | 8, 9 | 746 | ||
BC143254.1 | - | 8, 9 | 703 | 9051756 | ||
BC144610.1 | - | 6, 7 | 494 | 9053141 | ||
BX647673.1 | - | 15, 16 | 2974 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
