siRNA Details
Gene Symbol: UTP14C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021645.5 | NP_067677.4 | 2 | 2989 | ||
GenBank mRNA | BC089407.1 | - | 1 | 2398 | 3065365 | |
BC122536.1 | - | 2 | 2766 | 40036054 | ||
D87455.1 | - | 2 | 2989 |
Gene Symbol: UTP14A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166221.1 | NP_001159693.1 | 14 | 2193 | ||
NM_006649.3 | NP_006640.2 | 15 | 2349 | |||
XM_017029237.1 | XP_016884726.1 | 17 | 2501 | |||
XM_017029238.1 | XP_016884727.1 | 15 | 2531 | |||
GenBank mRNA | AK291918.1 | - | 15 | 2316 | ||
AK298578.1 | - | 14 | 2162 | |||
AK304563.1 | - | 14 | 2214 | |||
BC001149.1 | - | 15 | 2285 | 3355662 | ||
BC009649.1 | - | 15 | 2287 | 3459192 | ||
BC014987.1 | - | 15 | 2277 | 4901477 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
