siRNA Details
Gene Symbol: UTP14A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166221.1 | NP_001159693.1 | 11 | 1630 | ||
NM_006649.3 | NP_006640.2 | 12 | 1786 | |||
XM_017029237.1 | XP_016884726.1 | 14 | 1938 | |||
XM_017029238.1 | XP_016884727.1 | 12 | 1968 | |||
GenBank mRNA | AK291918.1 | - | 12 | 1753 | ||
AK298578.1 | - | 11 | 1599 | |||
AK304563.1 | - | 11 | 1651 | |||
BC001149.1 | - | 12 | 1722 | 3355662 | ||
BC009649.1 | - | 12 | 1724 | 3459192 | ||
BC014987.1 | - | 12 | 1714 | 4901477 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
