siRNA Details
Gene Symbol: TRMU | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282782.1 | NP_001269711.1 | 3 | 720 | ||
NM_001282783.1 | NP_001269712.1 | 3 | 720 | |||
NM_001282784.1 | NP_001269713.1 | 3 | 720 | |||
NM_001282785.1 | NP_001269714.1 | 4 | 827 | |||
NM_018006.4 | NP_060476.2 | 4 | 827 | |||
NR_104240.1 | 4 | 827 | ||||
NR_104241.1 | 3 | 720 | ||||
XM_005261678.1 | XP_005261735.1 | 4 | 514 | |||
XM_005261681.1 | XP_005261738.1 | 4 | 514 | |||
XM_011530271.2 | XP_011528573.1 | 4 | 590 | |||
XM_011530272.1 | XP_011528574.1 | 4 | 514 | |||
XM_011530273.1 | XP_011528575.1 | 4 | 514 | |||
XM_011530274.2 | XP_011528576.1 | 3 | 427 | |||
XM_011530275.1 | XP_011528577.1 | 4 | 470 | |||
GenBank mRNA | AB178028.1 | - | 4 | 514 | ||
AK001002.1 | - | 4 | 514 | |||
AK290712.1 | - | 4 | 507 | |||
AK295173.1 | - | 4 | 514 | |||
AK303020.1 | - | 4 | 566 | |||
AY062123.2 | - | 4 | 803 | |||
BC027991.1 | - | 3 | 378 | 5000797 | ||
BC080631.1 | - | 3 | 382 | 6536541 | ||
BX952612.1 | - | 3 | 348 | |||
CN365141.1 | - | 4 | 476 | |||
CR456445.1 | - | 4 | 483 | |||
DQ309998.1 | - | 3 | 696 | |||
DQ309999.1 | - | 3 | 696 | |||
DQ310000.1 | - | 3 | 696 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
