siRNA Details
Gene Symbol: PLPP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032728.3 | NP_116117.3 | 2 | 1564 | ||
GenBank mRNA | AK027568.1 | - | 2 | 1518 | ||
AK075207.1 | - | 2 | 1564 | |||
BC006362.2 | - | 2 | 1521 | 4129897 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
