siRNA Details
Gene Symbol: NUP35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287584.1 | NP_001274513.1 | 3 | 357 | ||
NM_001287585.1 | NP_001274514.1 | 2 | 254 | |||
NM_138285.4 | NP_612142.2 | 2 | 254 | |||
NR_109856.1 | 2 | 254 | ||||
XM_006712254.2 | XP_006712317.1 | 2 | 219 | |||
XM_011510576.2 | XP_011508878.1 | 4 | 485 | |||
XM_011510577.2 | XP_011508879.1 | 4 | 672 | |||
XM_017003308.1 | XP_016858797.1 | 2 | 410 | |||
GenBank mRNA | AF411516.1 | - | 2 | 254 | ||
AF514993.1 | - | 2 | 211 | |||
AK289601.1 | - | 2 | 197 | |||
AK290760.1 | - | 2 | 166 | |||
AK298199.1 | - | 2 | 164 | |||
AK302350.1 | - | 3 | 326 | |||
BC035597.1 | - | 2 | 178 | |||
BC047029.1 | - | 2 | 241 | 5541023 | ||
BC061698.1 | - | 2 | 208 | 4132440 | ||
BI818393.1 | - | 2 | 233 | |||
BI821231.1 | - | 2 | 190 | |||
CX787176.1 | - | 2 | 152 |

More Information
Other Species Targeted

Gene Ontology Categories:
