siRNA Details
Gene Symbol: NUP35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287584.1 | NP_001274513.1 | 3 | 362 | ||
NM_001287585.1 | NP_001274514.1 | 2 | 259 | |||
NM_138285.4 | NP_612142.2 | 2 | 259 | |||
NR_109856.1 | 2 | 259 | ||||
XM_006712254.2 | XP_006712317.1 | 2 | 224 | |||
XM_011510576.2 | XP_011508878.1 | 4 | 490 | |||
XM_011510577.2 | XP_011508879.1 | 4 | 677 | |||
XM_017003308.1 | XP_016858797.1 | 2 | 415 | |||
GenBank mRNA | AF411516.1 | - | 2 | 259 | ||
AF514993.1 | - | 2 | 216 | |||
AK289601.1 | - | 2 | 202 | |||
AK290760.1 | - | 2 | 171 | |||
AK298199.1 | - | 2 | 169 | |||
AK302350.1 | - | 3 | 331 | |||
BC035597.1 | - | 2 | 183 | |||
BC047029.1 | - | 2 | 246 | 5541023 | ||
BC061698.1 | - | 2 | 213 | 4132440 | ||
BI818393.1 | - | 2 | 238 | |||
BI821231.1 | - | 2 | 195 | |||
CX787176.1 | - | 2 | 157 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
