siRNA Details
Gene Symbol: TTYH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005367.2 | NP_001005367.1 | 13 | 1804 | ||
NM_001201461.1 | NP_001188390.1 | 14 | 1853 | |||
NM_020659.3 | NP_065710.1 | 14 | 1850 | |||
XM_017027025.1 | XP_016882514.1 | 13 | 1648 | |||
XM_017027026.1 | XP_016882515.1 | 14 | 1694 | |||
GenBank mRNA | AB162930.1 | - | 13 | 1686 | ||
AB188496.1 | - | 14 | 1853 | |||
AF177909.1 | - | 14 | 1763 | |||
BC011347.1 | - | 13 | 1709 | 4215136 | ||
BC019358.1 | - | 11 | 1186 | 3960435 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
