siRNA Details
Gene Symbol: UBB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281716.1 | NP_001268645.1 | 2 | 1070 | ||
NM_001281717.1 | NP_001268646.1 | 2 | 1041 | |||
NM_001281718.1 | NP_001268647.1 | 2 | 1001 | |||
NM_001281719.1 | NP_001268648.1 | 2 | 999 | |||
NM_001281720.1 | NP_001268649.1 | 2 | 845 | |||
NM_018955.3 | NP_061828.1 | 2 | 1214 | |||
GenBank mRNA | AK300882.1 | - | 3 | 692 | ||
BC000379.2 | - | 2 | 958 | 2820408 | ||
BC009301.2 | - | 2 | 904 | 4126622 | ||
BC015127.1 | - | 2 | 920 | 3929901 | ||
BC026301.1 | - | 2 | 916 | 4280629 | ||
BC038999.1 | - | 2 | 927 | 4943285 | ||
BC046123.2 | - | 2 | 906 | 3879581 | ||
GQ472200.1 | - | 2 | 958 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
