siRNA Details
Gene Symbol: UBB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281716.1 | NP_001268645.1 | 2 | 414 | ||
NM_001281717.1 | NP_001268646.1 | 2 | 385 | |||
NM_001281718.1 | NP_001268647.1 | 2 | 345 | |||
NM_001281719.1 | NP_001268648.1 | 2 | 343 | |||
NM_001281720.1 | NP_001268649.1 | 2 | 189 | |||
NM_018955.3 | NP_061828.1 | 2 | 558 | |||
GenBank mRNA | AK129498.1 | - | 2 | 260 | ||
AK300882.1 | - | 2 | 264 | |||
BC009301.2 | - | 2 | 248 | 4126622 | ||
BC015127.1 | - | 2 | 264 | 3929901 | ||
BC026301.1 | - | 2 | 260 | 4280629 | ||
BC031027.1 | - | 2 | 262 | 4715377 | ||
BC038999.1 | - | 2 | 271 | 4943285 | ||
BC046123.2 | - | 2 | 250 | 3879581 | ||
BG548604.1 | - | 2 | 346 | |||
BG718912.1 | - | 2 | 352 | |||
BM554254.1 | - | 2 | 268 | |||
BT020104.1 | - | 1 | 164 | |||
BU596366.1 | - | 2 | 363 | |||
BU661443.1 | - | 2 | 203 | |||
DB480830.1 | - | 2 | 385 | |||
HY014043.1 | - | 2 | 415 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
