siRNA Details
Gene Symbol: EGLN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053046.3 | NP_444274.1 | 3 | 1334 | ||
NM_080732.3 | NP_542770.2 | 3 | 1244 | |||
GenBank mRNA | AJ310544.1 | - | 2 | 1186 | ||
AK026863.1 | - | 1 | 280 | |||
AK074408.1 | - | 3 | 1173 | |||
AK098182.1 | - | 2 | 1918 | |||
AK314741.1 | - | 1 | 602 | |||
AL133009.1 | - | 1 | 2529 | |||
AL832506.1 | - | 3 | 1200 | |||
AY040565.1 | - | 2 | 1178 | |||
BC001723.1 | - | 3 | 1147 | |||
BC036051.1 | - | 3 | 1248 | 5270945 |
Gene Symbol: RAB4B-EGLN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037791.1 | 9 | 1938 | |||
GenBank mRNA | AK291385.1 | - | 9 | 1886 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
