siRNA Details
Gene Symbol: ZNF32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005368.2 | NP_001005368.1 | 2 | 335 | ||
NM_001324164.1 | NP_001311093.1 | 2 | 197 | |||
NM_001324165.1 | NP_001311094.1 | 2 | 335 | |||
NM_001324166.1 | NP_001311095.1 | 2 | 197 | |||
NM_001324167.1 | NP_001311096.1 | 2 | 197 | |||
NM_001324168.1 | NP_001311097.1 | 2 | 335 | |||
NM_006973.2 | NP_008904.1 | 2 | 197 | |||
XM_017016616.1 | XP_016872105.1 | 1 | 3268 | |||
GenBank mRNA | AI871265.1 | - | 2 | 240 | ||
AK309483.1 | - | 2 | 204 | |||
AK312132.1 | - | 2 | 194 | |||
BC022842.1 | - | 1 | 132 | 5014207 | ||
BG477527.1 | - | 2 | 199 | |||
HY045180.1 | - | 2 | 336 | |||
HY148951.1 | - | 2 | 319 | |||
U69645.1 | - | 2 | 173 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
