siRNA Details
Gene Symbol: GCSAM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190259.1 | NP_001177188.1 | 6 | 2774 | ||
NM_001190260.1 | NP_001177189.1 | 5 | 2723 | |||
NM_152785.4 | NP_689998.1 | 6 | 2768 | |||
XM_005247272.2 | XP_005247329.1 | 6 | 3192 | |||
XM_011512627.2 | XP_011510929.1 | 6 | 2646 | |||
XM_011512628.2 | XP_011510930.1 | 6 | 2703 | |||
XM_011512629.1 | XP_011510931.1 | 6 | 3195 | |||
XM_011512630.1 | XP_011510932.1 | 6 | 3201 | |||
GenBank mRNA | AY212246.1 | - | 6 | 2688 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
