siRNA Details
Gene Symbol: GCSHP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033248.1 | 1 | 529 | |||
GenBank mRNA | AK092810.1 | - | 2 | 2413 | ||
BM685716.1 | - | 1 | 282 |
Gene Symbol: GCSH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004483.4 | NP_004474.2 | 5 | 685 | ||
NR_033249.1 | 4 | 621 | ||||
XM_017023136.1 | XP_016878625.1 | 5 | 702 | |||
XM_017023137.1 | XP_016878626.1 | 5 | 645 | |||
GenBank mRNA | BC000790.1 | - | 1 | 609 | 3451361 | |
BC009065.1 | - | 1 | 644 | |||
BC020922.1 | - | 1 | 648 | 4247157 | ||
BF798008.1 | - | 5 | 556 | |||
BG708388.1 | - | 5 | 718 | |||
CN283805.1 | - | 4 | 509 | |||
M69175.1 | - | 5 | 612 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
