siRNA Details
Gene Symbol: GCSH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004483.4 | NP_004474.2 | 5 | 960 | ||
NR_033249.1 | 4 | 896 | ||||
XM_017023136.1 | XP_016878625.1 | 5 | 977 | |||
XM_017023137.1 | XP_016878626.1 | 5 | 920 | |||
GenBank mRNA | BC000790.1 | - | 1 | 884 | 3451361 | |
BC009065.1 | - | 1 | 919 | |||
BC020922.1 | - | 1 | 923 | 4247157 | ||
D00723.1 | - | 1 | 954 | |||
M69175.1 | - | 5 | 887 |
Gene Symbol: LOC729080 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033244.1 | 1 | 865 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
