siRNA Details
Gene Symbol: CD200 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004196.3 | NP_001004196.2 | 7 | 1357 | ||
NM_001318826.1 | NP_001305755.1 | 6 | 1277 | |||
NM_001318828.1 | NP_001305757.1 | 5 | 1200 | |||
NM_001318830.1 | NP_001305759.1 | 5 | 1195 | |||
NM_005944.6 | NP_005935.4 | 6 | 1282 | |||
XM_017006454.1 | XP_016861943.1 | 7 | 1847 | |||
XM_017006455.1 | XP_016861944.1 | 6 | 1482 | |||
XM_017006456.1 | XP_016861945.1 | 7 | 1208 | |||
XM_017006458.1 | XP_016861947.1 | 7 | 1848 | |||
XM_017006459.1 | XP_016861948.1 | 6 | 1467 | |||
GenBank mRNA | AF063591.1 | - | 6 | 1073 | ||
AK074972.1 | - | 6 | 1047 | |||
AK129797.1 | - | 5 | 964 | |||
AK293399.1 | - | 5 | 969 | |||
AK297194.1 | - | 7 | 1148 | |||
AL832529.1 | - | 6 | 1067 | |||
BC022522.1 | - | 6 | 1075 | 4794050 | ||
BC031103.1 | - | 6 | 1051 | 5301702 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
