siRNA Details
Gene Symbol: OR13C2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004481.1 | NP_001004481.1 | 1 | 483 | ||
GenBank mRNA | BC136824.1 | - | 1 | 526 | 9020815 | |
BC136826.1 | - | 1 | 526 | 9020817 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
